Assay Details
Gene Symbol: LOC5574180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661187.2 | XP_001661237.1 | 1 - 2 | 649 | 69 | |
GenBank mRNA | DQ440474.1 | - | 1 - 2 | 119 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
