Assay Details
Gene Symbol: LOC5574446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661413.2 | XP_001661463.1 | 1 - 2 | 213 | 74 | |
GenBank mRNA | DQ459304.1 | - | 1 - 2 | 123 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
