Assay Details
Gene Symbol: LOC5574446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661413.2 | XP_001661463.1 | 4 - 5 | 956 | 68 | |
GenBank mRNA | DQ459304.1 | - | 4 - 5 | 866 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
