Assay Details
Gene Symbol: LOC5580248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844522.1 | XP_021700214.1 | 5 - 6 | 796 | 76 | |
XM_021844523.1 | XP_021700215.1 | 6 - 7 | 901 | 76 | ||
GenBank mRNA | DQ459411.1 | - | 4 - 5 | 604 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
