Assay Details
Gene Symbol: LOC5580214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852862.1 | XP_021708554.1 | 2 - 3 | 1016 | 66 | |
XM_021852863.1 | XP_021708555.1 | 2 - 3 | 1063 | 66 | ||
XM_021852864.1 | XP_021708556.1 | 2 - 3 | 1018 | 66 | ||
XM_021852865.1 | XP_021708557.1 | 2 - 3 | 1017 | 66 | ||
XM_021852866.1 | XP_021708558.1 | 2 - 3 | 1014 | 66 | ||
XM_021852867.1 | XP_021708559.1 | 2 - 3 | 1057 | 66 | ||
XM_021852868.1 | XP_021708560.1 | 2 - 3 | 1062 | 66 | ||
XM_021852869.1 | XP_021708561.1 | 2 - 3 | 1016 | 66 | ||
XM_021852870.1 | XP_021708562.1 | 2 - 3 | 1060 | 66 | ||
GenBank mRNA | EF127524.1 | - | 1 - 2 | 156 | 66 | |
EF127525.1 | - | 1 - 2 | 156 | 66 | ||
EF127526.1 | - | 1 - 2 | 156 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
