Assay Details
Gene Symbol: LOC5580214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852862.1 | XP_021708554.1 | 3 - 4 | 115 | ||
XM_021852863.1 | XP_021708555.1 | 3 - 4 | 115 | |||
XM_021852864.1 | XP_021708556.1 | 3 - 4 | 115 | |||
XM_021852865.1 | XP_021708557.1 | 3 - 4 | 115 | |||
XM_021852866.1 | XP_021708558.1 | 3 - 4 | 115 | |||
XM_021852867.1 | XP_021708559.1 | 3 - 4 | 115 | |||
XM_021852868.1 | XP_021708560.1 | 3 - 4 | 115 | |||
XM_021852869.1 | XP_021708561.1 | 3 - 4 | 115 | |||
XM_021852870.1 | XP_021708562.1 | 3 - 4 | 115 | |||
GenBank mRNA | EF127524.1 | - | 2 - 3 | 394 | 115 | |
EF127525.1 | - | 2 - 3 | 394 | 115 | ||
EF127526.1 | - | 2 - 3 | 394 | 115 |

More Information
Set Membership: |
![]() ![]() |
