Assay Details
Gene Symbol: LOC23687644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845472.1 | XP_021701164.1 | 4 - 5 | 3156 | 70 | |
XM_021845473.1 | XP_021701165.1 | 4 - 5 | 2906 | 70 | ||
XM_021845474.1 | XP_021701166.1 | 4 - 5 | 3163 | 70 | ||
XM_021845475.1 | XP_021701167.1 | 4 - 5 | 3153 | 70 | ||
GenBank mRNA | EF153765.1 | - | 2 - 3 | 399 | 70 | |
EF153766.1 | - | 2 - 3 | 399 | 70 | ||
EF153767.1 | - | 2 - 3 | 399 | 70 | ||
EF153768.1 | - | 2 - 3 | 399 | 70 | ||
EF153769.1 | - | 2 - 3 | 399 | 70 | ||
EF153770.1 | - | 2 - 3 | 399 | 70 |

More Information
Set Membership: |
![]() ![]() |
