Assay Details
Gene Symbol: LOC23687644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845472.1 | XP_021701164.1 | 7 - 8 | 3741 | 96 | |
XM_021845473.1 | XP_021701165.1 | 7 - 8 | 3491 | 96 | ||
XM_021845474.1 | XP_021701166.1 | 7 - 8 | 3748 | 96 | ||
XM_021845475.1 | XP_021701167.1 | 7 - 8 | 3738 | 96 | ||
GenBank mRNA | EF153765.1 | - | 5 - 6 | 984 | 96 | |
EF153766.1 | - | 5 - 6 | 984 | 96 | ||
EF153767.1 | - | 5 - 6 | 984 | 96 | ||
EF153770.1 | - | 5 - 6 | 984 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
