Assay Details
Gene Symbol: LOC5572256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839335.1 | XP_021695027.1 | 3 - 4 | 977 | 90 | |
XM_021839336.1 | XP_021695028.1 | 3 - 4 | 1012 | 90 | ||
XM_021839337.1 | XP_021695029.1 | 3 - 4 | 775 | 90 | ||
XM_021839338.1 | XP_021695030.1 | 2 - 3 | 1173 | 90 | ||
XM_021839339.1 | XP_021695031.1 | 2 - 3 | 1172 | 90 | ||
XM_021839340.1 | XP_021695032.1 | 3 - 4 | 1010 | 90 | ||
XM_021839341.1 | XP_021695033.1 | 2 - 3 | 1172 | 90 | ||
XM_021839342.1 | XP_021695034.1 | 2 - 3 | 1172 | 90 | ||
GenBank mRNA | EF175868.1 | - | 3 - 4 | 968 | 90 |
