Assay Details
Gene Symbol: LOC5572256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839335.1 | XP_021695027.1 | 8 - 9 | 3034 | 94 | |
XM_021839336.1 | XP_021695028.1 | 8 - 9 | 3069 | 94 | ||
XM_021839337.1 | XP_021695029.1 | 8 - 9 | 2832 | 94 | ||
XM_021839338.1 | XP_021695030.1 | 7 - 8 | 3227 | 94 | ||
GenBank mRNA | EF175868.1 | - | 8 - 9 | 3025 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
