Assay Details
Gene Symbol: LOC5578456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656927.3 | XP_001656977.3 | 3 - 4 | 931 | 71 | |
XM_021851331.1 | XP_021707023.1 | 5 - 6 | 2062 | 71 | ||
XM_021851332.1 | XP_021707024.1 | 5 - 6 | 2728 | 71 | ||
XM_021851333.1 | XP_021707025.1 | 5 - 6 | 2723 | 71 | ||
XM_021851334.1 | XP_021707026.1 | 5 - 6 | 2057 | 71 | ||
XM_021851336.1 | XP_021707028.1 | 5 - 6 | 1611 | 71 | ||
XM_021851337.1 | XP_021707029.1 | 5 - 6 | 1218 | 71 | ||
XM_021851338.1 | XP_021707030.1 | 4 - 5 | 1465 | 71 | ||
XM_021851339.1 | XP_021707031.1 | 5 - 6 | 2039 | 71 | ||
GenBank mRNA | EF209048.1 | - | 4 - 5 | 1382 | 71 |
