Assay Details
Gene Symbol: LOC5578456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656927.3 | XP_001656977.3 | 4 - 5 | 1120 | 69 | |
XM_021851331.1 | XP_021707023.1 | 6 - 7 | 2251 | 69 | ||
XM_021851332.1 | XP_021707024.1 | 6 - 7 | 2917 | 69 | ||
XM_021851333.1 | XP_021707025.1 | 6 - 7 | 2912 | 69 | ||
XM_021851334.1 | XP_021707026.1 | 6 - 7 | 2246 | 69 | ||
XM_021851336.1 | XP_021707028.1 | 6 - 7 | 1800 | 69 | ||
XM_021851337.1 | XP_021707029.1 | 6 - 7 | 1407 | 69 | ||
XM_021851338.1 | XP_021707030.1 | 5 - 6 | 1654 | 69 | ||
XM_021851339.1 | XP_021707031.1 | 6 - 7 | 2228 | 69 | ||
GenBank mRNA | EF209048.1 | - | 5 - 6 | 1571 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
