Assay Details
Gene Symbol: LOC5572572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654047.2 | XP_001654097.2 | 2 - 3 | 2860 | 69 | |
XM_021857349.1 | XP_021713041.1 | 4 - 5 | 3142 | 69 | ||
XM_021857350.1 | XP_021713042.1 | 4 - 5 | 3026 | 69 | ||
XM_021857351.1 | XP_021713043.1 | 4 - 5 | 2899 | 69 | ||
XM_021857352.1 | XP_021713044.1 | 4 - 5 | 2332 | 69 | ||
XM_021857353.1 | XP_021713045.1 | 4 - 5 | 2267 | 69 | ||
XM_021857355.1 | XP_021713047.1 | 4 - 5 | 2126 | 69 | ||
XM_021857356.1 | XP_021713048.1 | 4 - 5 | 2621 | 69 | ||
XM_021857357.1 | XP_021713049.1 | 4 - 5 | 2596 | 69 | ||
XM_021857358.1 | XP_021713050.1 | 4 - 5 | 2190 | 69 | ||
XM_021857359.1 | XP_021713051.1 | 4 - 5 | 2109 | 69 | ||
XM_021857360.1 | XP_021713052.1 | 4 - 5 | 2137 | 69 | ||
XM_021857361.1 | XP_021713053.1 | 4 - 5 | 2096 | 69 | ||
GenBank mRNA | EF537041.1 | - | 1 - 2 | 1349 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
