Assay Details
Gene Symbol: SPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848571.1 | XP_021704263.1 | 2 - 3 | 1889 | 66 | |
GenBank mRNA | EU106874.1 | - | 1 - 2 | 606 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
