Assay Details
Gene Symbol: SPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848571.1 | XP_021704263.1 | 3 - 4 | 2013 | 62 | |
GenBank mRNA | EU106874.1 | - | 2 - 3 | 730 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
