Assay Details
Gene Symbol: SPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848571.1 | XP_021704263.1 | 4 - 5 | 2078 | 64 | |
GenBank mRNA | EU106874.1 | - | 3 - 4 | 795 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
