Assay Details
Gene Symbol: SPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848571.1 | XP_021704263.1 | 5 - 6 | 2207 | 68 | |
GenBank mRNA | EU106874.1 | - | 4 - 5 | 924 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
