Assay Details
Gene Symbol: LOC5578675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855388.1 | XP_021711080.1 | 2 - 3 | 663 | 84 | |
GenBank mRNA | EU267006.1 | - | 1 - 2 | 139 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
