Assay Details
Gene Symbol: LOC5578675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855388.1 | XP_021711080.1 | 5 - 6 | 1351 | 67 | |
GenBank mRNA | EU267006.1 | - | 4 - 5 | 812 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
