Assay Details
Gene Symbol: LOC5578675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855388.1 | XP_021711080.1 | 7 - 8 | 2339 | 91 | |
GenBank mRNA | EU267006.1 | - | 6 - 7 | 1800 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
