Assay Details
Gene Symbol: LOC5578664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851370.1 | XP_021707062.1 | 4 - 5 | 1092 | 76 | |
GenBank mRNA | EU267008.1 | - | 4 - 5 | 964 | 76 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5578664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851370.1 | XP_021707062.1 | 4 - 5 | 1092 | 76 | |
GenBank mRNA | EU267008.1 | - | 4 - 5 | 964 | 76 |