Assay Details
Gene Symbol: LOC5578663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855164.1 | XP_021710856.1 | 3 - 4 | 1076 | 105 | |
XM_021855165.1 | XP_021710857.1 | 3 - 4 | 1032 | 105 | ||
XM_021855166.1 | XP_021710858.1 | 3 - 4 | 1013 | 105 | ||
XM_021855167.1 | XP_021710859.1 | 3 - 4 | 969 | 105 | ||
GenBank mRNA | EU267011.1 | - | 2 - 3 | 773 | 105 |

More Information
Set Membership: |
![]() ![]() |
