Assay Details
Gene Symbol: LOC5578663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855164.1 | XP_021710856.1 | 4 - 5 | 1324 | 92 | |
XM_021855165.1 | XP_021710857.1 | 4 - 5 | 1280 | 92 | ||
XM_021855166.1 | XP_021710858.1 | 4 - 5 | 1261 | 92 | ||
XM_021855167.1 | XP_021710859.1 | 4 - 5 | 1217 | 92 | ||
GenBank mRNA | EU267011.1 | - | 3 - 4 | 1021 | 92 |
