Assay Details
Gene Symbol: LOC5567558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847282.1 | XP_021702974.1 | 4 - 5 | 97 | ||
XM_021847289.1 | XP_021702981.1 | 4 - 5 | 97 | |||
XM_021847300.1 | XP_021702992.1 | 4 - 5 | 97 | |||
GenBank mRNA | EU604099.1 | - | 3 - 4 | 780 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
