Assay Details
Gene Symbol: LOC5567562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847260.1 | XP_021702952.1 | 2 - 3 | 1210 | 75 | |
GenBank mRNA | EU604100.1 | - | 2 - 3 | 605 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
