Assay Details
Gene Symbol: LOC5567572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847310.1 | XP_021703002.1 | 2 - 3 | 946 | 89 | |
GenBank mRNA | EU604101.1 | - | 1 - 2 | 498 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
