Assay Details
Gene Symbol: LOC5570234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658963.2 | XP_001659013.1 | 1 - 2 | 287 | 76 | |
XM_021855794.1 | XP_021711486.1 | 2 - 3 | 243 | 76 | ||
GenBank mRNA | FJ357570.1 | - | 1 - 2 | 50 | 76 |
