Assay Details
Gene Symbol: LOC5570234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658963.2 | XP_001659013.1 | 3 - 4 | 566 | 82 | |
XM_021855794.1 | XP_021711486.1 | 4 - 5 | 522 | 82 | ||
GenBank mRNA | FJ357570.1 | - | 3 - 4 | 329 | 82 |
