Assay Details
Gene Symbol: LOC23687671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494700.2 | XP_011493002.2 | 1 - 2 | 1005 | 69 | |
GenBank mRNA | FJ979880.1 | - | 2 - 3 | 966 | 69 |

More Information
Set Membership: |
![]() ![]() |
