Assay Details
Gene Symbol: LOC23687671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494700.2 | XP_011493002.2 | 3 - 4 | 2926 | 61 | |
GenBank mRNA | FJ979880.1 | - | 4 - 5 | 2887 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
