Assay Details
Gene Symbol: LOC5566108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839746.1 | XP_021695438.1 | 4 - 5 | 1170 | 93 | |
XM_021839747.1 | XP_021695439.1 | 4 - 5 | 1194 | 93 | ||
XM_021839748.1 | XP_021695440.1 | 4 - 5 | 1169 | 93 |
