Assay Details
Gene Symbol: LOC5566108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839746.1 | XP_021695438.1 | 6 - 7 | 1436 | 109 | |
XM_021839747.1 | XP_021695439.1 | 6 - 7 | 1460 | 109 | ||
XM_021839748.1 | XP_021695440.1 | 6 - 7 | 1435 | 109 |

More Information
Set Membership: |
![]() ![]() |
