Assay Details
Gene Symbol: LOC5566108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839745.1 | XP_021695437.1 | 12 - 13 | 3215 | 96 | |
XM_021839746.1 | XP_021695438.1 | 12 - 13 | 2977 | 96 | ||
XM_021839747.1 | XP_021695439.1 | 12 - 13 | 3001 | 96 | ||
XM_021839748.1 | XP_021695440.1 | 11 - 12 | 2853 | 96 | ||
XM_021839749.1 | XP_021695441.1 | 11 - 12 | 2797 | 96 | ||
XM_021839750.1 | XP_021695442.1 | 10 - 11 | 2712 | 96 |
