Assay Details
Gene Symbol: LOC5566108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839745.1 | XP_021695437.1 | 13 - 14 | 3561 | 105 | |
XM_021839746.1 | XP_021695438.1 | 13 - 14 | 3323 | 105 | ||
XM_021839747.1 | XP_021695439.1 | 13 - 14 | 3347 | 105 | ||
XM_021839748.1 | XP_021695440.1 | 12 - 13 | 3199 | 105 | ||
XM_021839749.1 | XP_021695441.1 | 12 - 13 | 3143 | 105 | ||
XM_021839750.1 | XP_021695442.1 | 11 - 12 | 3058 | 105 |

More Information
Set Membership: |
![]() ![]() |
