Assay Details
Gene Symbol: LOC5566108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839745.1 | XP_021695437.1 | 14 - 15 | 4284 | 95 | |
XM_021839746.1 | XP_021695438.1 | 14 - 15 | 4046 | 95 | ||
XM_021839747.1 | XP_021695439.1 | 14 - 15 | 4070 | 95 | ||
XM_021839748.1 | XP_021695440.1 | 13 - 14 | 3922 | 95 | ||
XM_021839749.1 | XP_021695441.1 | 13 - 14 | 3866 | 95 | ||
XM_021839750.1 | XP_021695442.1 | 12 - 13 | 3781 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
