Assay Details
Gene Symbol: LOC5567513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849339.1 | XP_021705031.1 | 4 - 5 | 1561 | 64 | |
GenBank mRNA | AF098717.1 | - | 4 - 5 | 1532 | 64 |

More Information
Set Membership: |
![]() ![]() |
