Assay Details
Gene Symbol: LOC5567513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849339.1 | XP_021705031.1 | 5 - 6 | 1887 | 61 | |
GenBank mRNA | AF098717.1 | - | 5 - 6 | 1858 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
