Assay Details
Gene Symbol: LOC5563590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001647881.2 | XP_001647931.1 | 2 - 3 | 363 | 79 | |
GenBank mRNA | AF154067.1 | - | 1 - 2 | 217 | 79 | |
CR938364.1 | - | 2 - 3 | 257 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
