Assay Details
Gene Symbol: LOC5574485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655096.2 | XP_001655146.1 | 2 - 3 | 246 | 68 | |
GenBank mRNA | AF315552.1 | - | 2 - 3 | 141 | 68 | |
AY133344.1 | - | 1 - 2 | 134 | 68 | ||
AY432624.1 | - | 2 - 3 | 198 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
