Assay Details
Gene Symbol: LOC5571188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653466.2 | XP_001653516.2 | 1 - 2 | 296 | 97 | |
GenBank mRNA | AF325508.1 | - | 1 - 2 | 221 | 97 | |
AY194224.1 | - | 1 - 2 | 103 | 97 | ||
AY194225.1 | - | 1 - 2 | 103 | 97 |
