Assay Details
Gene Symbol: LOC5571188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653466.2 | XP_001653516.2 | 5 - 6 | 1358 | 124 | |
GenBank mRNA | AF325508.1 | - | 5 - 6 | 1283 | 124 | |
AY194225.1 | - | 5 - 6 | 1165 | 124 |

More Information
Set Membership: |
![]() ![]() |
