Assay Details
Gene Symbol: LOC5570613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850734.1 | XP_021706426.1 | 4 - 5 | 799 | 76 | |
XM_021850739.1 | XP_021706431.1 | 4 - 5 | 801 | 76 | ||
GenBank mRNA | AF369383.1 | - | 4 - 5 | 530 | 76 | |
AF543193.1 | - | 4 - 5 | 602 | 76 | ||
AY330296.1 | - | 3 - 4 | 713 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
