Assay Details
Gene Symbol: LOC5570613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850734.1 | XP_021706426.1 | 5 - 6 | 1066 | 98 | |
GenBank mRNA | AF369383.1 | - | 5 - 6 | 797 | 98 | |
AF543193.1 | - | 5 - 6 | 869 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
