Assay Details
Gene Symbol: LOC5570613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850734.1 | XP_021706426.1 | 8 - 9 | 1434 | 85 | |
XM_021850739.1 | XP_021706431.1 | 8 - 9 | 1436 | 85 | ||
GenBank mRNA | AF369383.1 | - | 8 - 9 | 1165 | 85 | |
AF543193.1 | - | 8 - 9 | 1237 | 85 | ||
AY330296.1 | - | 7 - 8 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
