Assay Details
Gene Symbol: LOC5570613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850734.1 | XP_021706426.1 | 9 - 10 | 69 | ||
XM_021850739.1 | XP_021706431.1 | 9 - 10 | 69 | |||
GenBank mRNA | AF369383.1 | - | 9 - 10 | 1273 | 68 | |
AF543193.1 | - | 9 - 10 | 69 | |||
AY330296.1 | - | 8 - 9 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
