Assay Details
Gene Symbol: LOC5579480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841989.1 | XP_021697681.1 | 7 - 8 | 1634 | 58 | |
XM_021841990.1 | XP_021697682.1 | 7 - 8 | 1506 | 58 | ||
XM_021841992.1 | XP_021697684.1 | 7 - 8 | 1634 | 58 | ||
XM_021841993.1 | XP_021697685.1 | 7 - 8 | 1506 | 58 | ||
XM_021841994.1 | XP_021697686.1 | 7 - 8 | 1869 | 58 | ||
XM_021841995.1 | XP_021697687.1 | 6 - 7 | 982 | 58 | ||
XM_021841996.1 | XP_021697688.1 | 5 - 6 | 1119 | 58 | ||
XM_021841997.1 | XP_021697689.1 | 6 - 7 | 1090 | 58 | ||
XM_021841998.1 | XP_021697690.1 | 5 - 6 | 663 | 58 | ||
XM_021841999.1 | XP_021697691.1 | 6 - 7 | 1101 | 58 | ||
XM_021842000.1 | XP_021697692.1 | 6 - 7 | 1097 | 58 | ||
XM_021842001.1 | XP_021697693.1 | 5 - 6 | 982 | 58 | ||
GenBank mRNA | AF393728.1 | - | 1 - 2 | 120 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
