Assay Details
Gene Symbol: LOC5579480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841989.1 | XP_021697681.1 | 8 - 9 | 1717 | 89 | |
XM_021841990.1 | XP_021697682.1 | 8 - 9 | 1589 | 89 | ||
XM_021841992.1 | XP_021697684.1 | 8 - 9 | 1717 | 89 | ||
XM_021841993.1 | XP_021697685.1 | 8 - 9 | 1589 | 89 | ||
XM_021841994.1 | XP_021697686.1 | 8 - 9 | 1952 | 89 | ||
XM_021841995.1 | XP_021697687.1 | 7 - 8 | 1065 | 89 | ||
XM_021841996.1 | XP_021697688.1 | 6 - 7 | 1202 | 89 | ||
XM_021841997.1 | XP_021697689.1 | 7 - 8 | 1173 | 89 | ||
XM_021841998.1 | XP_021697690.1 | 6 - 7 | 746 | 89 | ||
XM_021841999.1 | XP_021697691.1 | 7 - 8 | 1184 | 89 | ||
XM_021842000.1 | XP_021697692.1 | 7 - 8 | 1180 | 89 | ||
XM_021842001.1 | XP_021697693.1 | 6 - 7 | 1065 | 89 | ||
GenBank mRNA | AF393728.1 | - | 2 - 3 | 203 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
