Assay Details
Gene Symbol: LOC5579480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841989.1 | XP_021697681.1 | 9 - 10 | 1893 | 61 | |
XM_021841990.1 | XP_021697682.1 | 9 - 10 | 1765 | 61 | ||
XM_021841995.1 | XP_021697687.1 | 8 - 9 | 1241 | 61 | ||
XM_021841996.1 | XP_021697688.1 | 7 - 8 | 1378 | 61 | ||
XM_021841997.1 | XP_021697689.1 | 8 - 9 | 1349 | 61 | ||
XM_021841998.1 | XP_021697690.1 | 7 - 8 | 922 | 61 | ||
GenBank mRNA | AF393728.1 | - | 3 - 4 | 379 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
