Assay Details
Gene Symbol: LOC23687505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494791.2 | XP_011493093.2 | 1 - 2 | 230 | 83 | |
GenBank mRNA | AF393729.1 | - | 1 - 2 | 96 | 83 | |
AY432703.1 | - | 1 - 2 | 100 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011494791.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
