Assay Details
Gene Symbol: LOC23687505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494791.2 | XP_011493093.2 | 2 - 3 | 285 | 116 | |
GenBank mRNA | AF393729.1 | - | 2 - 3 | 151 | 116 | |
AY432703.1 | - | 2 - 3 | 155 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011494791.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
